Assay Details
Gene Symbol: med31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002417.2 | NP_001002417.2 | 3 - 4 | 340 | 114 | |
GenBank mRNA | BC076157.1 | - | 3 - 4 | 335 | 114 | |
EH553004.1 | - | 3 - 4 | 379 | 114 | ||
EH600668.1 | - | 1 - 2 | 530 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
