Assay Details
| Gene Symbol: rgs7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002541.1 | NP_001002541.1 | 15 - 16 | 1470 | 113 | |
| XM_009292967.3 | XP_009291242.1 | 16 - 17 | 1626 | 113 | ||
| XM_017351624.2 | XP_017207113.1 | 16 - 17 | 1624 | 113 | ||
| XM_021467140.1 | XP_021322815.1 | 16 - 17 | 1627 | 113 | ||
| XM_021467141.1 | XP_021322816.1 | 15 - 16 | 1412 | 113 | ||
| GenBank mRNA | AY817411.1 | - | 15 - 16 | 1417 | 113 | |
| BC076267.1 | - | 15 - 16 | 1470 | 113 | ||
More Information
Set Membership: |
|

rgs7a
Genomic Map