Assay Details
| Gene Symbol: lipia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003499.1 | NP_001003499.1 | 2 - 3 | 96 | 78 | |
| XM_021468811.1 | XP_021324486.1 | 3 - 4 | 396 | 78 | ||
| GenBank mRNA | BC078354.1 | - | 2 - 3 | 96 | 78 | |
More Information
Set Membership: |
|

lipia
Genomic Map