Assay Details
| Gene Symbol: lipia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003499.1 | NP_001003499.1 | 7 - 8 | 930 | 137 | |
| XM_021468811.1 | XP_021324486.1 | 8 - 9 | 1230 | 137 | ||
| GenBank mRNA | BC078354.1 | - | 7 - 8 | 930 | 137 | |
More Information
Set Membership: |
|

lipia
Genomic Map