Assay Details
| Gene Symbol: lipia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003499.1 | NP_001003499.1 | 9 - 10 | 1125 | 121 | |
| XM_021468811.1 | XP_021324486.1 | 10 - 11 | 1425 | 121 | ||
| GenBank mRNA | BC078354.1 | - | 9 - 10 | 1125 | 121 | |
More Information
Set Membership: |
|

lipia
Genomic Map