Assay Details
| Gene Symbol: cdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003983.1 | NP_001003983.1 | 11 - 12 | 1952 | 65 | |
| GenBank mRNA | AY496430.1 | - | 11 - 12 | 1952 | 65 | |
More Information
Set Membership: |
|

cdh5
Genomic Map