Assay Details
| Gene Symbol: myo6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004111.2 | NP_001004111.2 | 22 - 23 | 2410 | 76 | |
| XM_005160309.4 | XP_005160366.1 | 23 - 24 | 2556 | 76 | ||
| XM_005160311.4 | XP_005160368.1 | 23 - 24 | 2445 | 76 | ||
| XM_009294500.3 | XP_009292775.1 | 24 - 25 | 2565 | 76 | ||
| XM_009294501.3 | XP_009292776.1 | 24 - 25 | 2565 | 76 | ||
| XM_009294502.3 | XP_009292777.1 | 24 - 25 | 2565 | 76 | ||
| XM_021468402.1 | XP_021324077.1 | 24 - 25 | 2565 | 76 | ||
| XM_021468403.1 | XP_021324078.1 | 22 - 23 | 2404 | 76 | ||
| XM_021468404.1 | XP_021324079.1 | 23 - 24 | 2556 | 76 | ||
| GenBank mRNA | AY618702.1 | - | 22 - 23 | 2408 | 76 | |
| AY691329.1 | - | 23 - 24 | 2451 | 76 | ||
More Information
Set Membership: |
|

myo6a
Genomic Map