Assay Details
| Gene Symbol: myo6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004111.2 | NP_001004111.2 | 28 - 29 | 3141 | 71 | |
| XM_005160309.4 | XP_005160366.1 | 29 - 30 | 3287 | 71 | ||
| XM_009294500.3 | XP_009292775.1 | 30 - 31 | 3296 | 71 | ||
| XM_021468402.1 | XP_021324077.1 | 30 - 31 | 3296 | 71 | ||
| XM_021468403.1 | XP_021324078.1 | 28 - 29 | 3135 | 71 | ||
| XM_021468404.1 | XP_021324079.1 | 29 - 30 | 3287 | 71 | ||
| GenBank mRNA | AY618702.1 | - | 28 - 29 | 3139 | 71 | |
More Information
Set Membership: |
|

myo6a
Genomic Map