Assay Details
| Gene Symbol: myo6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004111.2 | NP_001004111.2 | 31 - 32 | 3422 | 70 | |
| XM_021468404.1 | XP_021324079.1 | 32 - 33 | 3568 | 70 | ||
| GenBank mRNA | AY618702.1 | - | 31 - 32 | 3420 | 70 | |
More Information
Set Membership: |
|

myo6a
Genomic Map