Assay Details
| Gene Symbol: myo6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004111.2 | NP_001004111.2 | 9 - 10 | 900 | 102 | |
| XM_005160309.4 | XP_005160366.1 | 10 - 11 | 1046 | 102 | ||
| XM_005160311.4 | XP_005160368.1 | 10 - 11 | 935 | 102 | ||
| XM_021468403.1 | XP_021324078.1 | 9 - 10 | 894 | 102 | ||
| XM_021468404.1 | XP_021324079.1 | 10 - 11 | 1046 | 102 | ||
| GenBank mRNA | AY618702.1 | - | 9 - 10 | 898 | 102 | |
| AY691329.1 | - | 10 - 11 | 941 | 102 | ||
More Information
Set Membership: |
|

myo6a
Genomic Map