Assay Details
Gene Symbol: fabp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004682.1 | NP_001004682.1 | 3 - 4 | 408 | 83 | |
GenBank mRNA | AY628221.2 | - | 3 - 4 | 411 | 83 | |
BC081489.1 | - | 3 - 4 | 408 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
