Assay Details
| Gene Symbol: dhdh.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005600.1 | NP_001005600.1 | 3 - 4 | 389 | 71 | |
| GenBank mRNA | BC081393.1 | - | 3 - 4 | 389 | 71 | |
| BC164754.1 | - | 3 - 4 | 405 | 71 | ||
More Information
Set Membership: |
|

dhdh.2
Genomic Map