Assay Details
| Gene Symbol: atp5pb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005960.1 | NP_001005960.1 | 5 - 6 | 584 | 87 | |
| GenBank mRNA | BC083308.1 | - | 5 - 6 | 584 | 87 | |
| BC165938.1 | - | 5 - 6 | 530 | 87 | ||
More Information
Set Membership: |
|

atp5pb
Genomic Map