Assay Details
| Gene Symbol: lbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007134.1 | NP_001007135.1 | 1 - 2 | 635 | 65 | |
| GenBank mRNA | BC075912.1 | - | 1 - 2 | 635 | 65 | |
More Information
Set Membership: |
|

lbx2
Genomic Map