Assay Details
Gene Symbol: bin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007421.1 | NP_001007422.1 | 6 - 7 | 567 | 105 | |
XM_009296976.3 | XP_009295251.1 | 6 - 7 | 661 | 105 | ||
XM_009296977.3 | XP_009295252.1 | 6 - 7 | 714 | 105 | ||
GenBank mRNA | BC085363.1 | - | 6 - 7 | 567 | 105 | |
BC165739.1 | - | 5 - 6 | 454 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
