Assay Details
| Gene Symbol: cnih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013524.1 | NP_001013542.1 | 1 - 2 | 381 | 58 | |
| XM_021479171.1 | XP_021334846.1 | 3 - 4 | 938 | 58 | ||
| XM_021479172.1 | XP_021334847.1 | 3 - 4 | 935 | 58 | ||
| GenBank mRNA | BC090830.1 | - | 1 - 2 | 381 | 58 | |
More Information
Set Membership: |
|

cnih2
Genomic Map