Assay Details
Gene Symbol: ppie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017678.1 | NP_001017678.1 | 7 - 8 | 529 | 62 | |
XM_009293090.3 | XP_009291365.3 | 7 - 8 | 564 | 62 | ||
GenBank mRNA | BC093329.1 | - | 7 - 8 | 529 | 62 | |
BC165323.1 | - | 7 - 8 | 546 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
