Assay Details
Gene Symbol: mtmr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020721.1 | NP_001018557.1 | 5 - 6 | 583 | 86 | |
XM_005172285.4 | XP_005172342.1 | 6 - 7 | 722 | 86 | ||
XM_005172286.4 | XP_005172343.1 | 5 - 6 | 656 | 86 | ||
XM_009303059.3 | XP_009301334.1 | 7 - 8 | 746 | 86 | ||
XM_009303060.3 | XP_009301335.1 | 6 - 7 | 680 | 86 | ||
GenBank mRNA | BC095884.1 | - | 5 - 6 | 583 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
