Assay Details
| Gene Symbol: endog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024214.1 | NP_001019385.1 | 2 - 3 | 766 | 91 | |
| GenBank mRNA | BC095134.1 | - | 2 - 3 | 766 | 91 | |
| BC164258.1 | - | 2 - 3 | 684 | 91 | ||
More Information
Set Membership: |
|

endog
Genomic Map