Assay Details
| Gene Symbol: polr2j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024400.1 | NP_001019571.1 | 2 - 3 | 182 | 92 | |
| GenBank mRNA | BC095123.1 | - | 2 - 3 | 182 | 92 | |
| BC165488.1 | - | 2 - 3 | 180 | 92 | ||
More Information
Set Membership: |
|

polr2j
Genomic Map