Assay Details
| Gene Symbol: coa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030084.1 | NP_001025255.1 | 2 - 3 | 518 | 83 | |
| GenBank mRNA | BC115052.1 | - | 2 - 3 | 501 | 83 | |
More Information
Set Membership: |
|

coa7
Genomic Map