Assay Details
| Gene Symbol: dnm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030128.1 | NP_001025299.1 | 13 - 14 | 1739 | 64 | |
| XM_005164243.4 | XP_005164300.1 | 13 - 14 | 1739 | 64 | ||
| XM_021470875.1 | XP_021326550.1 | 14 - 15 | 1783 | 64 | ||
| XM_021470883.1 | XP_021326558.1 | 13 - 14 | 1729 | 64 | ||
| GenBank mRNA | BC097134.1 | - | 13 - 14 | 1739 | 64 | |
| KC968470.1 | - | 13 - 14 | 1739 | 64 | ||
More Information
Set Membership: |
|

dnm2a
Genomic Map