Assay Details
| Gene Symbol: dnm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030128.1 | NP_001025299.1 | 17 - 18 | 2242 | 64 | |
| XM_005164241.4 | XP_005164298.1 | 18 - 19 | 2244 | 64 | ||
| XM_005164243.4 | XP_005164300.1 | 17 - 18 | 2242 | 64 | ||
| XM_021470873.1 | XP_021326548.1 | 19 - 20 | 2646 | 64 | ||
| XM_021470874.1 | XP_021326549.1 | 19 - 20 | 2298 | 64 | ||
| XM_021470875.1 | XP_021326550.1 | 18 - 19 | 2286 | 64 | ||
| XM_021470877.1 | XP_021326552.1 | 18 - 19 | 2265 | 64 | ||
| XM_021470881.1 | XP_021326556.1 | 18 - 19 | 2244 | 64 | ||
| XM_021470883.1 | XP_021326558.1 | 17 - 18 | 2232 | 64 | ||
| GenBank mRNA | BC097134.1 | - | 17 - 18 | 2242 | 64 | |
| KC968470.1 | - | 17 - 18 | 2242 | 64 | ||
More Information
Set Membership: |
|

dnm2a
Genomic Map