Assay Details
| Gene Symbol: daam1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030136.2 | NP_001025307.1 | 15 - 16 | 2006 | 74 | |
| XM_005160502.4 | XP_005160559.1 | 16 - 17 | 2213 | 74 | ||
| XM_005160503.4 | XP_005160560.1 | 16 - 17 | 2407 | 74 | ||
More Information
Set Membership: |
|

daam1b
Genomic Map