Assay Details
| Gene Symbol: daam1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030136.2 | NP_001025307.1 | 24 - 25 | 3027 | 85 | |
| XM_005160502.4 | XP_005160559.1 | 25 - 26 | 3234 | 85 | ||
| XM_005160503.4 | XP_005160560.1 | 25 - 26 | 3428 | 85 | ||
| XM_017352491.2 | XP_017207980.1 | 24 - 25 | 3398 | 85 | ||
| GenBank mRNA | BC163491.1 | - | 23 - 24 | 3030 | 85 | |
More Information
Set Membership: |
|

daam1b
Genomic Map