Assay Details
| Gene Symbol: nde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030203.1 | NP_001025374.1 | 3 - 4 | 463 | 107 | |
| GenBank mRNA | BC097169.1 | - | 3 - 4 | 463 | 107 | |
More Information
Set Membership: |
|

nde1
Genomic Map