Assay Details
| Gene Symbol: pacsin1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033728.1 | NP_001028900.1 | 3 - 4 | 323 | 74 | |
| XM_005166140.3 | XP_005166197.1 | 3 - 4 | 417 | 74 | ||
| XM_009302728.3 | XP_009301003.1 | 3 - 4 | 640 | 74 | ||
| GenBank mRNA | BC081593.1 | - | 3 - 4 | 378 | 74 | |
| BC097107.1 | - | 3 - 4 | 323 | 74 | ||
| BC165817.1 | - | 2 - 3 | 251 | 74 | ||
More Information
Set Membership: |
|

pacsin1b
Genomic Map