Assay Details
| Gene Symbol: pcyt1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037374.1 | NP_001032451.1 | 2 - 3 | 269 | 97 | |
| XM_005157481.4 | XP_005157538.1 | 2 - 3 | 309 | 97 | ||
| GenBank mRNA | BC109442.1 | - | 2 - 3 | 304 | 97 | |
More Information
Set Membership: |
|

pcyt1bb
Genomic Map