Assay Details
| Gene Symbol: inppl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039804.2 | NP_001034893.1 | 19 - 20 | 2922 | 64 | |
| GenBank mRNA | DQ272661.2 | - | 19 - 20 | 2922 | 64 | |
More Information
Set Membership: |
|

inppl1a
Genomic Map