Assay Details
| Gene Symbol: inppl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039804.2 | NP_001034893.1 | 9 - 10 | 1769 | 58 | |
| GenBank mRNA | BC171582.1 | - | 10 - 11 | 1689 | 58 | |
| DQ272661.2 | - | 9 - 10 | 1769 | 58 | ||
More Information
Set Membership: |
|

inppl1a
Genomic Map