Assay Details
Gene Symbol: pdlim3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042718.1 | NP_001036183.1 | 2 - 3 | 253 | 82 | |
XM_005157234.4 | XP_005157291.1 | 2 - 3 | 414 | 82 | ||
XM_005157236.4 | XP_005157293.1 | 2 - 3 | 414 | 82 | ||
GenBank mRNA | BC163828.1 | - | 2 - 3 | 305 | 82 | |
BC163837.1 | - | 2 - 3 | 305 | 82 | ||
DQ679228.1 | - | 2 - 3 | 253 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
