Assay Details
| Gene Symbol: fzd3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042761.2 | NP_001036226.1 | 3 - 4 | 862 | 62 | |
| XM_005160748.4 | XP_005160805.1 | 3 - 4 | 914 | 62 | ||
| GenBank mRNA | AB246776.1 | - | 2 - 3 | 387 | 62 | |
| BC161660.1 | - | 3 - 4 | 846 | 62 | ||
| BC163439.1 | - | 2 - 3 | 517 | 62 | ||
| BC163452.1 | - | 2 - 3 | 517 | 62 | ||
| DQ200953.1 | - | 3 - 4 | 862 | 62 | ||
More Information
Set Membership: |
|

fzd3a
Genomic Map