Assay Details
| Gene Symbol: fzd3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042761.2 | NP_001036226.1 | 4 - 5 | 1886 | 79 | |
| XM_005160748.4 | XP_005160805.1 | 4 - 5 | 1938 | 79 | ||
| GenBank mRNA | AB246776.1 | - | 3 - 4 | 1411 | 79 | |
| BC163439.1 | - | 3 - 4 | 1541 | 79 | ||
| BC163452.1 | - | 3 - 4 | 1541 | 79 | ||
| DQ200953.1 | - | 4 - 5 | 1886 | 79 | ||
More Information
Set Membership: |
|

fzd3a
Genomic Map