Assay Details
| Gene Symbol: loxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043325.1 | NP_001036790.1 | 2 - 3 | 966 | 71 | |
| GenBank mRNA | DQ304647.1 | - | 2 - 3 | 966 | 71 | |
| EF030480.1 | - | 2 - 3 | 954 | 71 | ||
More Information
Set Membership: |
|

loxl1
Genomic Map