Assay Details
| Gene Symbol: pycr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044938.1 | NP_001038403.1 | 4 - 5 | 422 | 78 | |
| GenBank mRNA | BC124544.1 | - | 4 - 5 | 346 | 78 | |
More Information
Set Membership: |
|

pycr3
Genomic Map