Assay Details
| Gene Symbol: hpse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045005.1 | NP_001038470.1 | 11 - 12 | 1664 | 61 | |
| GenBank mRNA | BC139574.1 | - | 11 - 12 | 1657 | 61 | |
More Information
Set Membership: |
|

hpse
Genomic Map