Assay Details
| Gene Symbol: rpain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045388.1 | NP_001038853.1 | 2 - 3 | 324 | 60 | |
| XM_005171732.4 | XP_005171789.1 | 2 - 3 | 377 | 60 | ||
| GenBank mRNA | BC122206.1 | - | 2 - 3 | 324 | 60 | |
More Information
Set Membership: |
|

rpain
Genomic Map