Assay Details
| Gene Symbol: abhd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079677.1 | NP_001073145.1 | 3 - 4 | 599 | 80 | |
| XM_005163017.2 | XP_005163074.1 | 5 - 6 | 808 | 80 | ||
| XM_009297903.3 | XP_009296178.1 | 4 - 5 | 675 | 80 | ||
| GenBank mRNA | BC124444.1 | - | 3 - 4 | 599 | 80 | |
| BC165844.1 | - | 3 - 4 | 593 | 80 | ||
More Information
Set Membership: |
|

abhd2b
Genomic Map