Assay Details
| Gene Symbol: abhd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079677.1 | NP_001073145.1 | 8 - 9 | 1129 | 82 | |
| XM_005163017.2 | XP_005163074.1 | 10 - 11 | 1338 | 82 | ||
| XM_009297903.3 | XP_009296178.1 | 9 - 10 | 1205 | 82 | ||
| GenBank mRNA | BC124444.1 | - | 8 - 9 | 1129 | 82 | |
| BC165844.1 | - | 8 - 9 | 1123 | 82 | ||
More Information
Set Membership: |
|

abhd2b
Genomic Map