Assay Details
Gene Symbol: arhgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082868.1 | NP_001076337.1 | 5 - 6 | 536 | 55 | |
XM_005161867.4 | XP_005161924.1 | 7 - 8 | 989 | 55 | ||
XM_005161868.4 | XP_005161925.3 | 6 - 7 | 1970 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
