Assay Details
| Gene Symbol: npffr1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089389.1 | NP_001082858.1 | 3 - 4 | 577 | 85 | |
| GenBank mRNA | BC134148.1 | - | 3 - 4 | 577 | 85 | |
| GU290221.1 | - | 3 - 4 | 594 | 85 | ||
More Information
Set Membership: |
|

npffr1l3
Genomic Map