Assay Details
| Gene Symbol: tmigd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098248.1 | NP_001091718.1 | 4 - 5 | 725 | 97 | |
| GenBank mRNA | CU458924.1 | - | 4 - 5 | 725 | 97 | |
More Information
Set Membership: |
|

tmigd1
Genomic Map