Assay Details
Gene Symbol: slc9a6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098256.2 | NP_001091726.2 | 13 - 14 | 1728 | 84 | |
XM_009291141.3 | XP_009289416.1 | 14 - 15 | 1873 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
