Assay Details
Gene Symbol: puf60a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105588.1 | NP_001099058.1 | 9 - 10 | 1059 | 64 | |
XM_005163509.4 | XP_005163566.1 | 8 - 9 | 1001 | 64 | ||
XM_021480690.1 | XP_021336365.1 | 8 - 9 | 1001 | 64 | ||
GenBank mRNA | BC135083.1 | - | 9 - 10 | 1071 | 64 | |
BC153301.1 | - | 9 - 10 | 1059 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
