Assay Details
Gene Symbol: trpc4apa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131569.2 | NP_571644.1 | 16 - 17 | 1874 | 66 | |
GenBank mRNA | AF130459.1 | - | 16 - 17 | 1875 | 66 | |
BC099990.1 | - | 5 - 6 | 484 | 66 | ||
BC115197.1 | - | 5 - 6 | 498 | 66 | ||
BC134876.1 | - | 5 - 6 | 514 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
