Assay Details
Gene Symbol: trpc4apa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131569.2 | NP_571644.1 | 17 - 18 | 1988 | 96 | |
GenBank mRNA | AF130459.1 | - | 17 - 18 | 1989 | 96 | |
BC099990.1 | - | 6 - 7 | 598 | 96 | ||
BC115197.1 | - | 6 - 7 | 612 | 96 | ||
BC134876.1 | - | 6 - 7 | 628 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
