Assay Details
| Gene Symbol: dap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131572.2 | NP_571647.1 | 3 - 4 | 318 | 95 | |
| GenBank mRNA | AF231127.1 | - | 3 - 4 | 195 | 95 | |
| BC049052.1 | - | 3 - 4 | 318 | 95 | ||
More Information
Set Membership: |
|

dap
Genomic Map