Assay Details
| Gene Symbol: cry3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB042251.1 | - | 12 - 13 | 1748 | 54 | |
| CF995949.1 | - | 3 - 4 | 212 | 54 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cry3b| Gene Symbol: cry3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB042251.1 | - | 12 - 13 | 1748 | 54 | |
| CF995949.1 | - | 3 - 4 | 212 | 54 | ||
Set Membership: |
|