Assay Details
| Gene Symbol: cry3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131792.2 | NP_571867.2 | 14 - 15 | 1989 | 56 | |
| XM_009295768.2 | XP_009294043.2 | 14 - 15 | 2724 | 56 | ||
| GenBank mRNA | AB042251.1 | - | 13 - 14 | 1872 | 56 | |
| CF995949.1 | - | 4 - 5 | 336 | 56 | ||
More Information
Set Membership: |
|

cry3b
Genomic Map