Assay Details
| Gene Symbol: cry3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131792.2 | NP_571867.2 | 10 - 11 | 1616 | 71 | |
| XM_009295768.2 | XP_009294043.2 | 10 - 11 | 2348 | 71 | ||
| XM_021469382.1 | XP_021325057.1 | 10 - 11 | 2350 | 71 | ||
| GenBank mRNA | AB042251.1 | - | 9 - 10 | 1499 | 71 | |
More Information
Set Membership: |
|

cry3b
Genomic Map